missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-D-phenylalanine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2168914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Fmoc-D-phenylalanine | |
181°C to 185°C | |
MFCD00062955 | |
fmoc-d-phenylalanine, fmoc-d-phe-oh, n-fmoc-d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-d-phenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid, fmoc-r-beta-phenylalanine, n-9-fluorenylmethoxycarbonyl-d-phenylalanine, d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-phenylalanine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-phenyl-propanoic acid | |
+40° (c=1 in DMF) | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid | |
978332 | |
98% |
86123-10-6 | |
C24H21NO4 | |
25 g | |
SJVFAHZPLIXNDH-JOCHJYFZSA-N | |
OC(=O)[C@@H](CC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
387.44 | |
387.44 |
Chemical Identifiers
86123-10-6 | |
387.44 | |
SJVFAHZPLIXNDH-JOCHJYFZSA-N | |
978332 | |
OC(=O)[C@@H](CC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
C24H21NO4 | |
MFCD00062955 | |
fmoc-d-phenylalanine, fmoc-d-phe-oh, n-fmoc-d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-d-phenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid, fmoc-r-beta-phenylalanine, n-9-fluorenylmethoxycarbonyl-d-phenylalanine, d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-phenylalanine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-phenyl-propanoic acid | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only