Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-norleucine, 95%

Chemical Identifiers

Chemical Name or Material N-Fmoc-D-norleucine
CAS 112883-41-7
Formula Weight 353.42
MDL Number MFCD00155639
Beilstein 8227505
Synonym Fmoc-D-Nle-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6672806

  SDS
alfa aesar™
H6672806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6672814

  SDS
alfa aesar™
H6672814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 353.42
Chemical Name or Material N-Fmoc-D-norleucine
CAS 112883-41-7
MDL Number MFCD00155639
Assay 95%
Color White
Storage Note 1 2° to 8°C
Physical Form Powder
Beilstein 8227505
Quantity 5g
Synonym Fmoc-D-Nle-OH
Safety and Handling

2° to 8°C