missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-methionine, 98%

$33.47 - $481.14

Chemical Identifiers

CAS 112883-40-6
Molecular Formula C20H21NO4S
Molecular Weight (g/mol) 371.451
MDL Number MFCD00062958
InChI Key BUBGAUHBELNDEW-GOSISDBHSA-N
Synonym fmoc-d-met-oh, fmoc-d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl-d-methionine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylthio butanoic acid, d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, n-fmoc-d-methionine, ambotzfaa1145, n-9-fluorenylmethyloxycarbonyl-d-methionine, fmoc-d-met-oh
PubChem CID 6992521
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfanylbutanoic acid
SMILES CSCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6217806 View Documents Alfa Aesar™
H6217806
5g Each for $33.47
 
AAH6217814 View Documents Alfa Aesar™
H6217814
25g Each for $132.62
 
AAH6217822 View Documents Alfa Aesar™
H6217822
100g Each for $481.14
 
Specifications

Chemical Identifiers

112883-40-6
371.451
BUBGAUHBELNDEW-GOSISDBHSA-N
6992521
CSCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C20H21NO4S
MFCD00062958
fmoc-d-met-oh, fmoc-d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl-d-methionine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylthio butanoic acid, d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, n-fmoc-d-methionine, ambotzfaa1145, n-9-fluorenylmethyloxycarbonyl-d-methionine, fmoc-d-met-oh
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfanylbutanoic acid

Specifications

112883-40-6
MFCD00062958
BUBGAUHBELNDEW-GOSISDBHSA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfanylbutanoic acid
6992521
98%
5g
N-Fmoc-D-methionine
C20H21NO4S
fmoc-d-met-oh, fmoc-d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl-d-methionine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylthio butanoic acid, d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, n-fmoc-d-methionine, ambotzfaa1145, n-9-fluorenylmethyloxycarbonyl-d-methionine, fmoc-d-met-oh
CSCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
371.451
371.45
130°C to 134°C
+30° (c=1 in DMF)