Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-homophenylalanine, 98%

Chemical Identifiers

Chemical Name or Material N-Fmoc-D-homophenylalanine
CAS 135944-09-1
Formula Weight 401.46
MDL Number MFCD00151917
Beilstein 4847668
Synonym Fmoc-D-HomoPhe-OH; (R)-2-(Fmoc-amino)-4-phenylbutyric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6674606

 
alfa aesar™
H6674606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6674603

 
alfa aesar™
H6674603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 401.46
Chemical Name or Material N-Fmoc-D-homophenylalanine
Physical Form Powder
CAS 135944-09-1
Synonym Fmoc-D-HomoPhe-OH; (R)-2-(Fmoc-amino)-4-phenylbutyric acid
MDL Number MFCD00151917
Color White
Quantity 5g
Storage Note 1 2° to 8°C
Beilstein 4847668
Assay 98%
Safety and Handling

2° to 8°C