missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-glutamine, 95%

$153.65 - $700.99

Chemical Identifiers

CAS 112898-00-7
Molecular Formula C20H20N2O5
Molecular Weight (g/mol) 368.389
MDL Number MFCD00062959
InChI Key IZKGGDFLLNVXNZ-QGZVFWFLSA-N
Synonym 2r-5-amino-2-9h-fluoren-9-ylmethoxy oxo methyl amino-5-oxopentanoic acid, ambotzfaa1483, fmoc-d-gln, fmoc-d-gln-oh, fmoc-d-glutamine, n-a-fmoc-d-glutamine, n-alpha-9-fluorenylmethoxycarbonyl-d-glutamine, n-alpha-9-fluorenylmethyloxycarbonyl-d-glutamine, n-alpha-fmoc-d-glutamine
PubChem CID 6992536
IUPAC Name (2R)-5-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxopentanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)N)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6609506
View Documents
Alfa Aesar™
H6609506
5g Each for $153.65
 
AAH6609514
View Documents
Alfa Aesar™
H6609514
25g Each for $700.99
 
Specifications

Chemical Identifiers

112898-00-7
368.389
IZKGGDFLLNVXNZ-QGZVFWFLSA-N
6992536
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)N)C(=O)O
C20H20N2O5
MFCD00062959
2r-5-amino-2-9h-fluoren-9-ylmethoxy oxo methyl amino-5-oxopentanoic acid, ambotzfaa1483, fmoc-d-gln, fmoc-d-gln-oh, fmoc-d-glutamine, n-a-fmoc-d-glutamine, n-alpha-9-fluorenylmethoxycarbonyl-d-glutamine, n-alpha-9-fluorenylmethyloxycarbonyl-d-glutamine, n-alpha-fmoc-d-glutamine
(2R)-5-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxopentanoic acid

Specifications

112898-00-7
MFCD00062959
IZKGGDFLLNVXNZ-QGZVFWFLSA-N
(2R)-5-amino-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxopentanoic acid
6992536
Powder
White
N-Fmoc-D-glutamine
C20H20N2O5
2r-5-amino-2-9h-fluoren-9-ylmethoxy oxo methyl amino-5-oxopentanoic acid, ambotzfaa1483, fmoc-d-gln, fmoc-d-gln-oh, fmoc-d-glutamine, n-a-fmoc-d-glutamine, n-alpha-9-fluorenylmethoxycarbonyl-d-glutamine, n-alpha-9-fluorenylmethyloxycarbonyl-d-glutamine, n-alpha-fmoc-d-glutamine
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)N)C(=O)O
368.389
368.39
95%
5g
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold