missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-D-glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals
$119.32 - $438.08
Chemical Identifiers
CAS | 104091-08-9 |
---|---|
Molecular Formula | C24H27NO6 |
Molecular Weight (g/mol) | 425.48 |
MDL Number | MFCD00077055 |
InChI Key | OTKXCALUHMPIGM-UHFFFAOYNA-N |
Synonym | fmoc-d-glu otbu-oh, fmoc-d-glutamic acid 5-tert-butyl ester, fmoc-d-glu obut-oh, fmoc-d-glutamic acid gamma-tert-butyl ester, commercial, 2r-5-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, ambotzfaa1324, pubchem10015, fmoc-d-glu tbu-oh, fmoc-d-glu otbu-oh h |
PubChem CID | 7018822 |
IUPAC Name | (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid |
SMILES | CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6662406
|
Thermo Scientific Chemicals
H6662406 |
5 g |
Each for $119.32
|
|
|||||
AAH6662414
|
Thermo Scientific Chemicals
H6662414 |
25 g |
Each for $438.08
|
|
|||||
Description
It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
104091-08-9 | |
425.48 | |
OTKXCALUHMPIGM-UHFFFAOYNA-N | |
7018822 | |
CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
C24H27NO6 | |
MFCD00077055 | |
fmoc-d-glu otbu-oh, fmoc-d-glutamic acid 5-tert-butyl ester, fmoc-d-glu obut-oh, fmoc-d-glutamic acid gamma-tert-butyl ester, commercial, 2r-5-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, ambotzfaa1324, pubchem10015, fmoc-d-glu tbu-oh, fmoc-d-glu otbu-oh h | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid |
Specifications
83°C to 89°C | |
C24H27NO6 | |
fmoc-d-glu otbu-oh, fmoc-d-glutamic acid 5-tert-butyl ester, fmoc-d-glu obut-oh, fmoc-d-glutamic acid gamma-tert-butyl ester, commercial, 2r-5-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, ambotzfaa1324, pubchem10015, fmoc-d-glu tbu-oh, fmoc-d-glu otbu-oh h | |
OTKXCALUHMPIGM-UHFFFAOYNA-N | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid | |
7018822 | |
98% | |
Solid |
104091-08-9 | |
MFCD00077055 | |
Slightly soluble in water. | |
CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
425.48 | |
425.48 | |
5 g | |
N-Fmoc-D-glutamic acid 5-tert-butyl ester |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only