missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-glutamic acid 5-benzyl ester, 95%

$28.84 - $337.73

Chemical Identifiers

CAS 104091-11-4
Molecular Formula C27H25NO6
Molecular Weight (g/mol) 459.498
MDL Number MFCD00273449
InChI Key HJJURMMMGPQIQP-XMMPIXPASA-N
Synonym fmoc-d-glu obzl-oh, fmoc-d-glu bzl-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid, 2r-5-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, ambotzfaa1485, fmoc-d-glutamic acid, a-benzyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-d-glutamic-acid-gamma-benzyl ester, 2r-5-benzyloxy-2-9h-fluoren-9-ylmethoxycarbonylamino-5-oxo-pentanoic acid
PubChem CID 10837618
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66018MD View Documents Alfa Aesar™
H66018MD
250mg Each for $28.84
 
AAH6601803 View Documents Alfa Aesar™
H6601803
1g Each for $85.14
 
AAH6601806 View Documents Alfa Aesar™
H6601806
5g Each for $337.73
 
Specifications

Chemical Identifiers

104091-11-4
459.498
HJJURMMMGPQIQP-XMMPIXPASA-N
10837618
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C27H25NO6
MFCD00273449
fmoc-d-glu obzl-oh, fmoc-d-glu bzl-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid, 2r-5-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, ambotzfaa1485, fmoc-d-glutamic acid, a-benzyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-d-glutamic-acid-gamma-benzyl ester, 2r-5-benzyloxy-2-9h-fluoren-9-ylmethoxycarbonylamino-5-oxo-pentanoic acid
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid

Specifications

104091-11-4
MFCD00273449
HJJURMMMGPQIQP-XMMPIXPASA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid
10837618
Solid
250mg
C27H25NO6
fmoc-d-glu obzl-oh, fmoc-d-glu bzl-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid, 2r-5-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, ambotzfaa1485, fmoc-d-glutamic acid, a-benzyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-d-glutamic-acid-gamma-benzyl ester, 2r-5-benzyloxy-2-9h-fluoren-9-ylmethoxycarbonylamino-5-oxo-pentanoic acid
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
459.498
459.5
95%
N-Fmoc-D-glutamic acid 5-benzyl ester
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C