missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-aspartic acid 4-tert-butyl ester, 98%

$33.78 - $396.32

Chemical Identifiers

CAS 112883-39-3
Molecular Formula C23H25NO6
Molecular Weight (g/mol) 411.454
MDL Number MFCD00077050
InChI Key FODJWPHPWBKDON-LJQANCHMSA-N
Synonym fmoc-d-asp otbu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-d-asp obut-oh, fmoc-d-aspartic acid beta-tert-butyl ester, n-alpha-fmoc-d-aspartic acid beta-t-butyl ester, 2r-4-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-dasp otbu, ambotzfaa1309, fmoc-dasp otbu-oh, pubchem10009
PubChem CID 7018816
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62356MD View Documents Alfa Aesar™
H62356MD
250mg Each for $33.78
 
AAH6235603 View Documents Alfa Aesar™
H6235603
1g Each for $99.61
 
AAH6235606 View Documents Alfa Aesar™
H6235606
5g Each for $396.32
 
Specifications

Chemical Identifiers

112883-39-3
411.454
FODJWPHPWBKDON-LJQANCHMSA-N
7018816
CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C23H25NO6
MFCD00077050
fmoc-d-asp otbu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-d-asp obut-oh, fmoc-d-aspartic acid beta-tert-butyl ester, n-alpha-fmoc-d-aspartic acid beta-t-butyl ester, 2r-4-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-dasp otbu, ambotzfaa1309, fmoc-dasp otbu-oh, pubchem10009
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid

Specifications

250mg
C23H25NO6
fmoc-d-asp otbu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-d-asp obut-oh, fmoc-d-aspartic acid beta-tert-butyl ester, n-alpha-fmoc-d-aspartic acid beta-t-butyl ester, 2r-4-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-dasp otbu, ambotzfaa1309, fmoc-dasp otbu-oh, pubchem10009
CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
411.454
411.45
N-Fmoc-D-aspartic acid 4-tert-butyl ester
112883-39-3
MFCD00077050
FODJWPHPWBKDON-LJQANCHMSA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
7018816
98%