Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-aspartic acid 4-benzyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Fmoc-D-aspartic acid 4-benzyl ester
CAS 150009-58-8
Formula Weight 445.47
MDL Number MFCD00273445
Synonym Fmoc-D-Asp(OBzl)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6642203

 
alfa aesar™
H6642203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6642206

 
alfa aesar™
H6642206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66422MD

 
alfa aesar™
H66422MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 445.47
Assay 95%
CAS 150009-58-8
Physical Form Solid
MDL Number MFCD00273445
Synonym Fmoc-D-Asp(OBzl)-OH
Quantity 1g
Chemical Name or Material N-Fmoc-D-aspartic acid 4-benzyl ester
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C