missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-aspartic acid 4-allyl ester, 95%

Manufacturer:  Alfa Aesar™ H66349MD

 View more versions of this product

Catalog No. AAH66349MD



Specifications

Specifications

177609-12-0
MFCD01074688
FBNFRRNBFASDKS-LJQANCHMSA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid
10872947
Solid
250mg
C22H21NO6
2r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-oxo-4-prop-2-en-1-yloxy butanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxo-4-prop-2-en-1-yloxy butanoic acid, d-aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl-, 4-2-propenyl ester, fmoc-d-asp oall-oh, n-alpha-9-fluorenylmethyloxycarbonyl-d-aspartic acid beta allyl ester, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-allyloxy-4-oxobutanoic acid, r-2-9h-fluoren-9-yl methoxy carbonylamino-4-allyloxy-4-oxobutanoic acid
C=CCOC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
395.411
395.41
95%

Chemical Identifiers

177609-12-0
395.411
FBNFRRNBFASDKS-LJQANCHMSA-N
10872947
C=CCOC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C22H21NO6
MFCD01074688
2r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-oxo-4-prop-2-en-1-yloxy butanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxo-4-prop-2-en-1-yloxy butanoic acid, d-aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl-, 4-2-propenyl ester, fmoc-d-asp oall-oh, n-alpha-9-fluorenylmethyloxycarbonyl-d-aspartic acid beta allyl ester, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-allyloxy-4-oxobutanoic acid, r-2-9h-fluoren-9-yl methoxy carbonylamino-4-allyloxy-4-oxobutanoic acid
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

Documents