Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-aspartic acid 1-allyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Fmoc-D-aspartic acid 1-allyl ester
CAS 204246-17-3
Formula Weight 395.41
MDL Number MFCD01074689
Synonym Fmoc-D-Asp(Oall)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6621006

 
alfa aesar™
H6621006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66210MD

 
alfa aesar™
H66210MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6621003

 
alfa aesar™
H6621003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 395.41
Chemical Name or Material N-Fmoc-D-aspartic acid 1-allyl ester
Physical Form Crystalline Powder
Assay 95%
MDL Number MFCD01074689
CAS 204246-17-3
Synonym Fmoc-D-Asp(Oall)-OH
Color White
Quantity 5g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C