missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-alpha-phenylglycine, 98%

Manufacturer:  Alfa Aesar™ H6382306

 View more versions of this product

Catalog No. AAH6382306



Specifications

Specifications

N-Fmoc-D-alpha-phenylglycine
C23H19NO4
5309006
PCJHOCNJLMFYCV-OAQYLSRUSA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2-phenylacetic acid
2756108
Solid
5g
111524-95-9
MFCD00190891
2r-2-9h-fluoren-9-yl methoxy carbonyl amino-2-phenylacetic acid, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-2-phenyl-acetic acid, a-phenylglycine, fmoc-d-alpha-phenylglycine, fmoc-d-phenylglycine, fmoc-d-phg-oh, fmoc-l-, n-alpha-9-fluorenylmethyloxycarbonyl-d-phenylglycine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-2-phenylacetic acid, r-9h-fluoren-9-ylmethoxy carbonyl amino phenyl acetic acid, r-9h-fluoren-9-ylmethoxycarbonylamino-phenyl-acetic acid
C1=CC=C(C=C1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
373.408
373.4
98%

Chemical Identifiers

111524-95-9
373.408
PCJHOCNJLMFYCV-OAQYLSRUSA-N
2756108
C1=CC=C(C=C1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C23H19NO4
MFCD00190891
2r-2-9h-fluoren-9-yl methoxy carbonyl amino-2-phenylacetic acid, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-2-phenyl-acetic acid, a-phenylglycine, fmoc-d-alpha-phenylglycine, fmoc-d-phenylglycine, fmoc-d-phg-oh, fmoc-l-, n-alpha-9-fluorenylmethyloxycarbonyl-d-phenylglycine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-2-phenylacetic acid, r-9h-fluoren-9-ylmethoxy carbonyl amino phenyl acetic acid, r-9h-fluoren-9-ylmethoxycarbonylamino-phenyl-acetic acid
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2-phenylacetic acid
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

Documents