Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-4-fluoro-D-phenylalanine, 95%

Chemical Identifiers

Chemical Name or Material N-Fmoc-4-fluoro-D-phenylalanine
CAS 177966-64-2
Formula Weight 405.43
MDL Number MFCD00235900
Synonym Fmoc-D-Phe(4-F)-OH; (R)-2-(Boc-amino)-3-(4-fluorophenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6692403

 
alfa aesar™
H6692403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6692406

 
alfa aesar™
H6692406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 405.43
CAS 177966-64-2
Chemical Name or Material N-Fmoc-4-fluoro-D-phenylalanine
Assay 95%
Synonym Fmoc-D-Phe(4-F)-OH; (R)-2-(Boc-amino)-3-(4-fluorophenyl)propionic acid
Color White
Physical Form Powder
Quantity 1g
Storage Note 1 2° to 8°C
MDL Number MFCD00235900
Safety and Handling

2° to 8°C