missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-4-chloro-L-phenylalanine, 98%

Manufacturer:  Alfa Aesar™ H63448MD

 View more versions of this product

Catalog No. AAH63448MD



Specifications

Specifications

N-Fmoc-4-chloro-L-phenylalanine
C24H20ClNO4
9437838
CQPNKLNINBUUOM-QFIPXVFZSA-N
(2S)-3-(4-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
2734471
Solid
250mg
175453-08-4
MFCD00080281
fmoc-4-chloro-l-phenylalanine, s-n-fmoc-4-chlorophenylalanine, fmoc-phe 4-cl-oh, fmoc-l-4-chlorophenylalanine, fmoc-l-phe 4-cl-oh, fmoc-4-chloro-phe-oh, fmoc-l-4-chlorophe, 2s-3-4-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-chlorophenyl propanoic acid, phenylalanine, 4-chloro-n-9h-fluoren-9-ylmethoxy carbonyl
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Cl)C(=O)O
421.877
421.87
98%

Chemical Identifiers

175453-08-4
421.877
CQPNKLNINBUUOM-QFIPXVFZSA-N
2734471
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Cl)C(=O)O
C24H20ClNO4
MFCD00080281
fmoc-4-chloro-l-phenylalanine, s-n-fmoc-4-chlorophenylalanine, fmoc-phe 4-cl-oh, fmoc-l-4-chlorophenylalanine, fmoc-l-phe 4-cl-oh, fmoc-4-chloro-phe-oh, fmoc-l-4-chlorophe, 2s-3-4-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-chlorophenyl propanoic acid, phenylalanine, 4-chloro-n-9h-fluoren-9-ylmethoxy carbonyl
(2S)-3-(4-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

Documents