missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-4-chloro-D-phenylalanine, 98%

Manufacturer:  Alfa Aesar™ H6359103

 View more versions of this product

Catalog No. AAH6359103



Specifications

Specifications

N-Fmoc-4-chloro-D-phenylalanine
C24H20ClNO4
8726004
CQPNKLNINBUUOM-JOCHJYFZSA-N
(2R)-3-(4-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
16213163
Powder
White
142994-19-2
MFCD00077066
fmoc-4-chloro-d-phenylalanine, fmoc-d-phe 4-cl-oh, fmoc-d-4-chlorophe, fmoc-d-4-chlorophenylalanine, fmoc-d-pclphe-oh, fmoc-p-chloro-d-phe-oh, fmoc-4-chloro-d-phe-oh, fmoc-pcl-d-phe-oh, d-phenylalanine, 4-chloro-n-9h-fluoren-9-ylmethoxy carbonyl, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-chlorophenyl propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Cl)C(=O)O
421.877
421.87
98%
1g

Chemical Identifiers

142994-19-2
421.877
CQPNKLNINBUUOM-JOCHJYFZSA-N
16213163
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Cl)C(=O)O
C24H20ClNO4
MFCD00077066
fmoc-4-chloro-d-phenylalanine, fmoc-d-phe 4-cl-oh, fmoc-d-4-chlorophe, fmoc-d-4-chlorophenylalanine, fmoc-d-pclphe-oh, fmoc-p-chloro-d-phe-oh, fmoc-4-chloro-d-phe-oh, fmoc-pcl-d-phe-oh, d-phenylalanine, 4-chloro-n-9h-fluoren-9-ylmethoxy carbonyl, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-chlorophenyl propanoic acid
(2R)-3-(4-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents