Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-4-chloro-D-phenylalanine, 98%

$31.24 - $31.24

Chemical Identifiers

Chemical Name or Material N-Fmoc-4-chloro-D-phenylalanine
CAS 142994-19-2
Formula Weight 421.87
MDL Number MFCD00077066
Beilstein 8726004
Synonym Fmoc-D-Phe(4-Cl)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6359103

 
alfa aesar™
H6359103
1g Each for $31.24

AAH6359106

 
alfa aesar™
H6359106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6359114

 
alfa aesar™
H6359114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 421.87
Chemical Name or Material N-Fmoc-4-chloro-D-phenylalanine
Physical Form Powder
Assay 98%
MDL Number MFCD00077066
CAS 142994-19-2
Beilstein 8726004
Synonym Fmoc-D-Phe(4-Cl)-OH
Color White
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C