Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-4-benzoyl-L-phenylalanine, 95%

$227.53 - $227.53

Chemical Identifiers

Chemical Name or Material N-Fmoc-4-benzoyl-L-phenylalanine
CAS 117666-96-3
Formula Weight 491.54
MDL Number MFCD00151937
Beilstein 7672022
Synonym Fmoc-Bpa-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6670703

 
alfa aesar™
H6670703
1g Each for $227.53

AAH6670706

 
alfa aesar™
H6670706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66707MD

 
alfa aesar™
H66707MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 491.54
Chemical Name or Material N-Fmoc-4-benzoyl-L-phenylalanine
CAS 117666-96-3
Physical Form Powder
MDL Number MFCD00151937
Synonym Fmoc-Bpa-OH
Beilstein 7672022
Color White
Quantity 1g
Storage Note 1 2° to 8°C
Assay 95%
Safety and Handling

2° to 8°C