missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-3-nitro-D-phenylalanine, 95%, Thermo Scientific™
$61.73 - $761.93
Chemical Identifiers
CAS | 478183-71-0 |
---|---|
Molecular Formula | C24H20N2O6 |
Molecular Weight (g/mol) | 432.432 |
MDL Number | MFCD01317715 |
InChI Key | UDIZJKKIJYRJIN-JOCHJYFZSA-N |
Synonym | fmoc-3-nitro-d-phenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3-nitrophenyl propanoic acid, fmoc-d-3-nitrophenylalanine, fmoc-d-3-no2-phe-oh, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-3-nitrophenyl propanoic acid, fmoc-l-phe 3-no2-oh, fmoc-d-3-nitrophe, n-9h-fluoren-9-yl methoxy carbonyl-3-nitro-d-phenylalanine, 2r-2 9h-fluoren-9-yl methoxy carbonyl amino-3-3-nitrophenyl propanoic acid |
PubChem CID | 7006635 |
IUPAC Name | (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(3-nitrophenyl)propanoic acid |
SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC(=CC=C4)[N+](=O)[O-])C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH65010MD
|
Thermo Scientific Chemicals
H65010MD |
250 mg |
Each for $61.73
|
|
|||||
AAH6501003
|
Thermo Scientific Chemicals
H6501003 |
1 g |
Each for $187.49
|
|
|||||
AAH6501006
|
Thermo Scientific Chemicals
H6501006 |
5 g |
Each for $761.93
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
478183-71-0 | |
432.432 | |
UDIZJKKIJYRJIN-JOCHJYFZSA-N | |
7006635 | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC(=CC=C4)[N+](=O)[O-])C(=O)O |
C24H20N2O6 | |
MFCD01317715 | |
fmoc-3-nitro-d-phenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3-nitrophenyl propanoic acid, fmoc-d-3-nitrophenylalanine, fmoc-d-3-no2-phe-oh, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-3-nitrophenyl propanoic acid, fmoc-l-phe 3-no2-oh, fmoc-d-3-nitrophe, n-9h-fluoren-9-yl methoxy carbonyl-3-nitro-d-phenylalanine, 2r-2 9h-fluoren-9-yl methoxy carbonyl amino-3-3-nitrophenyl propanoic acid | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(3-nitrophenyl)propanoic acid |
Specifications
151°C to 157°C | |
White | |
MFCD01317715 | |
UDIZJKKIJYRJIN-JOCHJYFZSA-N | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(3-nitrophenyl)propanoic acid | |
7006635 | |
95% | |
Powder |
478183-71-0 | |
C24H20N2O6 | |
fmoc-3-nitro-d-phenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3-nitrophenyl propanoic acid, fmoc-d-3-nitrophenylalanine, fmoc-d-3-no2-phe-oh, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-3-nitrophenyl propanoic acid, fmoc-l-phe 3-no2-oh, fmoc-d-3-nitrophe, n-9h-fluoren-9-yl methoxy carbonyl-3-nitro-d-phenylalanine, 2r-2 9h-fluoren-9-yl methoxy carbonyl amino-3-3-nitrophenyl propanoic acid | |
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC(=CC=C4)[N+](=O)[O-])C(=O)O | |
432.432 | |
432.43 | |
250 mg | |
N-Fmoc-3-nitro-D-phenylalanine |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold