Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-3-cyclohexyl-D-alanine, 96%

$55.76 - $55.76

Chemical Identifiers

Chemical Name or Material N-Fmoc-3-cyclohexyl-D-alanine
CAS 144701-25-7
Formula Weight 393.48
MDL Number MFCD00151912
Beilstein 8234680
Synonym Fmoc-D-Cha-OH; (R)-2-(Fmoc-amino)-3-cyclohexylpropionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6238203

  SDS
alfa aesar™
H6238203
1g Each for $55.76

AAH6238206

  SDS
alfa aesar™
H6238206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 393.48
Synonym Fmoc-D-Cha-OH; (R)-2-(Fmoc-amino)-3-cyclohexylpropionic acid
Assay 96%
Chemical Name or Material N-Fmoc-3-cyclohexyl-D-alanine
Color White
Quantity 1g
Storage Note 1 2° to 8°C
CAS 144701-25-7
MDL Number MFCD00151912
Beilstein 8234680
Physical Form Powder

Applications
N-Fmoc-3-cyclohexyl-D-alanine is used as fine chemical intermediate.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

2° to 8°C