Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-3,5-diiodo-L-tyrosine, 98%

Chemical Identifiers

Chemical Name or Material N-Fmoc-3,5-diiodo-L-tyrosine
CAS 103213-31-6
Formula Weight 655.22
MDL Number MFCD00063330
Beilstein 9022736
Synonym Fmoc-Tyr(3,5-I2)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6358003

  SDS
alfa aesar™
H6358003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63580MD

  SDS
alfa aesar™
H63580MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 655.22
Chemical Name or Material N-Fmoc-3,5-diiodo-L-tyrosine
Physical Form Powder
Assay 98%
MDL Number MFCD00063330
CAS 103213-31-6
Beilstein 9022736
Synonym Fmoc-Tyr(3,5-I2)-OH
Color White
Quantity 1g
Sensitivity Light Sensitive
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C