missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-3,5-diiodo-L-tyrosine, 98%

Manufacturer:  Alfa Aesar™ H63580MD

 View more versions of this product

Catalog No. AAH63580MD



Specifications

Specifications

N-Fmoc-3,5-diiodo-L-tyrosine
C24H19I2NO5
9022736
IKNWCIROCRMKAY-NRFANRHFSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid
15700510
Powder
White
Light Sensitive
103213-31-6
MFCD00063330
fmoc-tyr 3,5-i2-oh, fmoc-3,5-diiodo-l-tyrosine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid, l-tyrosine, n-9h-fluoren-9-ylmethoxy carbonyl-3,5-diiodo, n-alpha-9-fluorenylmethyloxycarbonyl-l-3,5-diiodo-tyrosine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid, ambotzfaa1751, fmoc-3,5-diiodo-tyr-oh, n-fmoc-s-3,5-diiodotyrosine, fmoc-tyr 3,5-i2-oh hplc
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC(=C(C(=C4)I)O)I)C(=O)O
655.227
655.22
98%
250mg

Chemical Identifiers

103213-31-6
655.227
IKNWCIROCRMKAY-NRFANRHFSA-N
15700510
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC(=C(C(=C4)I)O)I)C(=O)O
C24H19I2NO5
MFCD00063330
fmoc-tyr 3,5-i2-oh, fmoc-3,5-diiodo-l-tyrosine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid, l-tyrosine, n-9h-fluoren-9-ylmethoxy carbonyl-3,5-diiodo, n-alpha-9-fluorenylmethyloxycarbonyl-l-3,5-diiodo-tyrosine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid, ambotzfaa1751, fmoc-3,5-diiodo-tyr-oh, n-fmoc-s-3,5-diiodotyrosine, fmoc-tyr 3,5-i2-oh hplc
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents