Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-3-(2-pyridyl)-L-alanine, 95%

Chemical Identifiers

Chemical Name or Material N-Fmoc-3-(2-pyridyl)-L-alanine
CAS 185379-40-2
Formula Weight 388.42
MDL Number MFCD00672564
Synonym Fmoc-3-(2-pyridyl)-L-Ala-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6652903

 
alfa aesar™
H6652903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66529MD

 
alfa aesar™
H66529MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 388.42
Chemical Name or Material N-Fmoc-3-(2-pyridyl)-L-alanine
CAS 185379-40-2
Assay 95%
MDL Number MFCD00672564
Synonym Fmoc-3-(2-pyridyl)-L-Ala-OH
Storage Note 1 2° to 8°C
Physical Form Powder
Color Pale brown
Quantity 1g
Safety and Handling

2° to 8°C