Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-2-methyl-D-phenylalanine, 95%

Chemical Identifiers

Chemical Name or Material N-Fmoc-2-methyl-D-phenylalanine
CAS 352351-63-4
Formula Weight 401.46
MDL Number MFCD00671404
Beilstein 9157614
Synonym Fmoc-D-2-Me-Phe-OH; (R)-2-(Fmoc-amino)-3-(2-methylphenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6518803

 
alfa aesar™
H6518803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6518806

 
alfa aesar™
H6518806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 401.46
Melting Point 110° to 116°C
Chemical Name or Material N-Fmoc-2-methyl-D-phenylalanine
Assay 95%
CAS 352351-63-4
MDL Number MFCD00671404
Color White
Beilstein 9157614
Quantity 1g
Storage Note 1 2° to 8°C
Physical Form Powder
Synonym Fmoc-D-2-Me-Phe-OH; (R)-2-(Fmoc-amino)-3-(2-methylphenyl)propionic acid
Safety and Handling

2° to 8°C