Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-2-fluoro-L-phenylalanine, 95%

Chemical Identifiers

Chemical Name or Material N-Fmoc-2-fluoro-L-phenylalanine
CAS 205526-26-7
Formula Weight 405.43
MDL Number MFCD00672552
Beilstein 9437460
Synonym Fmoc-Phe(2-F)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6675103

 
alfa aesar™
H6675103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6675106

 
alfa aesar™
H6675106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66751MD

 
alfa aesar™
H66751MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 405.43
MDL Number MFCD00672552
Chemical Name or Material N-Fmoc-2-fluoro-L-phenylalanine
Synonym Fmoc-Phe(2-F)-OH
Physical Form Powder
Color White
Assay 95%
Beilstein 9437460
Storage Note 1 2° to 8°C
Quantity 1g
CAS 205526-26-7
Safety and Handling

2° to 8°C