missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-2-cyclopentyl-L-glycine, 95%

$88.68 - $1,054.81

Chemical Identifiers

CAS 220497-61-0
Molecular Formula C22H23NO4
Molecular Weight (g/mol) 365.429
MDL Number MFCD01311751
InChI Key JGWHUIQSEKGLAQ-FQEVSTJZSA-N
Synonym fmoc-cpg-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclopentylacetic acid, fmoc-cyclopentyl-gly-oh, s-cyclopentyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid, fmoc-l-cpg-oh, ambotzfaa1360, fmoc-l-cyclopentyl-glycine, n-fmoc-alpha-cyclopentylglycine, fmoc-s-2-cyclopentylglycine, n-alpha-9-fluorenylmethyloxycarbonyl-l-beta-cyclopentylglycine
PubChem CID 7006544
IUPAC Name (2S)-2-cyclopentyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid
SMILES C1CCC(C1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66154MD View Documents Alfa Aesar™
H66154MD
250mg Each for $88.68
 
AAH6615403 View Documents Alfa Aesar™
H6615403
1g Each for $263.70
 
AAH6615406 View Documents Alfa Aesar™
H6615406
5g Each for $1,054.81
 
Specifications

Chemical Identifiers

220497-61-0
365.429
JGWHUIQSEKGLAQ-FQEVSTJZSA-N
7006544
C1CCC(C1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C22H23NO4
MFCD01311751
fmoc-cpg-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclopentylacetic acid, fmoc-cyclopentyl-gly-oh, s-cyclopentyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid, fmoc-l-cpg-oh, ambotzfaa1360, fmoc-l-cyclopentyl-glycine, n-fmoc-alpha-cyclopentylglycine, fmoc-s-2-cyclopentylglycine, n-alpha-9-fluorenylmethyloxycarbonyl-l-beta-cyclopentylglycine
(2S)-2-cyclopentyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid

Specifications

220497-61-0
MFCD01311751
JGWHUIQSEKGLAQ-FQEVSTJZSA-N
(2S)-2-cyclopentyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid
7006544
Powder
White
N-Fmoc-2-cyclopentyl-L-glycine
C22H23NO4
fmoc-cpg-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclopentylacetic acid, fmoc-cyclopentyl-gly-oh, s-cyclopentyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid, fmoc-l-cpg-oh, ambotzfaa1360, fmoc-l-cyclopentyl-glycine, n-fmoc-alpha-cyclopentylglycine, fmoc-s-2-cyclopentylglycine, n-alpha-9-fluorenylmethyloxycarbonyl-l-beta-cyclopentylglycine
C1CCC(C1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
365.429
379.46
95%
250mg
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C