missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-2-cyclohexyl-L-glycine, 98%

Manufacturer:  Alfa Aesar™ H6358903

 View more versions of this product

Catalog No. AAH6358903



Specifications

Specifications

N-Fmoc-2-cyclohexyl-L-glycine
C23H25NO4
fmoc-chg-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclohexylacetic acid, s-cyclohexyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid, fmoc-l-cyclohexylglycine, ambotzfaa1713, fmoc-cyclohexyl-gly-oh, bicr147, n-fmoc-2-cyclohexyl-l-glycine, s-alpha-fmoc-amino cyclohexaneacetic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-cyclohexylglycine
C1CCC(CC1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
379.456
379.45
98%
161321-36-4
MFCD00190878
BWQQGHPODCJZDB-NRFANRHFSA-N
(2S)-2-cyclohexyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid
6976353
Solid
1g

Chemical Identifiers

161321-36-4
379.456
BWQQGHPODCJZDB-NRFANRHFSA-N
6976353
C1CCC(CC1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C23H25NO4
MFCD00190878
fmoc-chg-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclohexylacetic acid, s-cyclohexyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid, fmoc-l-cyclohexylglycine, ambotzfaa1713, fmoc-cyclohexyl-gly-oh, bicr147, n-fmoc-2-cyclohexyl-l-glycine, s-alpha-fmoc-amino cyclohexaneacetic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-cyclohexylglycine
(2S)-2-cyclohexyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents