missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-2-cyclohexyl-L-glycine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H63589MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Fmoc-2-cyclohexyl-L-glycine | |
C23H25NO4 | |
fmoc-chg-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclohexylacetic acid, s-cyclohexyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid, fmoc-l-cyclohexylglycine, ambotzfaa1713, fmoc-cyclohexyl-gly-oh, bicr147, n-fmoc-2-cyclohexyl-l-glycine, s-alpha-fmoc-amino cyclohexaneacetic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-cyclohexylglycine | |
OC(=O)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1CCCCC1 | |
379.46 | |
379.45 | |
250 mg |
161321-36-4 | |
MFCD00190878 | |
BWQQGHPODCJZDB-UHFFFAOYNA-N | |
2-cyclohexyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetic acid | |
6976353 | |
98% | |
Solid |
Chemical Identifiers
161321-36-4 | |
379.46 | |
BWQQGHPODCJZDB-UHFFFAOYNA-N | |
6976353 |
C23H25NO4 | |
MFCD00190878 | |
fmoc-chg-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclohexylacetic acid, s-cyclohexyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid, fmoc-l-cyclohexylglycine, ambotzfaa1713, fmoc-cyclohexyl-gly-oh, bicr147, n-fmoc-2-cyclohexyl-l-glycine, s-alpha-fmoc-amino cyclohexaneacetic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-cyclohexylglycine | |
OC(=O)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1CCCCC1 |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only