missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-2-cyclohexyl-D-glycine, 95%

$43.03 - $422.54

Chemical Identifiers

CAS 198543-96-3
Molecular Formula C23H25NO4
Molecular Weight (g/mol) 379.456
MDL Number MFCD00190877
InChI Key BWQQGHPODCJZDB-OAQYLSRUSA-N
Synonym fmoc-d-chg-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclohexylacetic acid, r-cyclohexyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid, ambotzfaa1666, bicr146, r-a-fmoc-amino-cyclohexaneacetic acid, r-alpha-fmoc-amino cyclohexaneacetic acid, n-alpha-9-fluorenylmethyloxycarbonyl-d-cyclohexylglycine, 2r-2-cyclohexyl-2-9h-fluoren-9-ylmethoxycarbonylamino acetic acid
PubChem CID 6976355
IUPAC Name (2R)-2-cyclohexyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid
SMILES C1CCC(CC1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66174MD View Documents Alfa Aesar™
H66174MD
250mg Each for $43.03
 
AAH6617403 View Documents Alfa Aesar™
H6617403
1g Each for $113.35
 
AAH6617406 View Documents Alfa Aesar™
H6617406
5g Each for $422.54
 
Specifications

Chemical Identifiers

198543-96-3
379.456
BWQQGHPODCJZDB-OAQYLSRUSA-N
6976355
C1CCC(CC1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C23H25NO4
MFCD00190877
fmoc-d-chg-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclohexylacetic acid, r-cyclohexyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid, ambotzfaa1666, bicr146, r-a-fmoc-amino-cyclohexaneacetic acid, r-alpha-fmoc-amino cyclohexaneacetic acid, n-alpha-9-fluorenylmethyloxycarbonyl-d-cyclohexylglycine, 2r-2-cyclohexyl-2-9h-fluoren-9-ylmethoxycarbonylamino acetic acid
(2R)-2-cyclohexyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid

Specifications

198543-96-3
MFCD00190877
BWQQGHPODCJZDB-OAQYLSRUSA-N
(2R)-2-cyclohexyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid
6976355
Solid
250mg
C23H25NO4
fmoc-d-chg-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclohexylacetic acid, r-cyclohexyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid, ambotzfaa1666, bicr146, r-a-fmoc-amino-cyclohexaneacetic acid, r-alpha-fmoc-amino cyclohexaneacetic acid, n-alpha-9-fluorenylmethyloxycarbonyl-d-cyclohexylglycine, 2r-2-cyclohexyl-2-9h-fluoren-9-ylmethoxycarbonylamino acetic acid
C1CCC(CC1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
379.456
379.46
95%
N-Fmoc-2-cyclohexyl-D-glycine
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C