missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-1-trityl-L-histidine, 98%

CAS: 109425-51-6 | C40H33N3O4 | 619.721 g/mol

Supplier:  Thermo Scientific Chemicals H5971114

 View more versions of this product

Catalog No. AAH5971114


Only null left
Explore available promotions

Description

Description

It is used in the synthesis and application of Fmoc-His(3-Bum)-OH.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis and application of Fmoc-His(3-Bum)-OH.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from heat. Keep it refrigerated.
Specifications

Specifications

N-Fmoc-1-trityl-L-histidine
145°C
MFCD00043332
5204720
fmoc-his trt-oh, ambotzfaa1090, nalpha-fmoc-nim-trityl-l-histidine, nalpha-fmoc-nim-trityl-l-histidine, n-a-fmoc-n-im-trityl-l-histidine, n, histidine, n-9h-fluoren-9-ylmethoxy carbonyl-1-triphenylmethyl, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-1-tritylimidazol-4-yl propanoic acid, fmochis trt oh, fmochis trt oh, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid
XXMYDXUIZKNHDT-QNGWXLTQSA-N
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57
619.721
619.71
109425-51-6
C40H33N3O4
25 g
Moisture sensitive
Slightly soluble in water.
+87° (c=5 in Chloroform)
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1-tritylimidazol-4-yl)propanoic acid
11422193
98%

Chemical Identifiers

109425-51-6
619.721
XXMYDXUIZKNHDT-QNGWXLTQSA-N
11422193
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57
C40H33N3O4
MFCD00043332
fmoc-his trt-oh, ambotzfaa1090, nalpha-fmoc-nim-trityl-l-histidine, nalpha-fmoc-nim-trityl-l-histidine, n-a-fmoc-n-im-trityl-l-histidine, n, histidine, n-9h-fluoren-9-ylmethoxy carbonyl-1-triphenylmethyl, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-1-tritylimidazol-4-yl propanoic acid, fmochis trt oh, fmochis trt oh, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1-tritylimidazol-4-yl)propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 439-640-4

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only