Please login to your online account to display your discounted pricing

N-Fluoropyridinium triflate, 95%, ACROS Organics™

$91.58 - $91.58

Chemical Identifiers

Chemical Name or Material N-Fluoropyridinium triflate
Name Note 95%
CAS 107263-95-6
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H5F4NO3S
Formula Weight 247.16
MDL Number MFCD00013458
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC259925000

  SDS
thermo fisher scientific
259925000
Glass bottle 500mg Each for $91.58
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder or Needles
Formula Weight 247.16
Quantity 500mg
Color White to off-white
Melting Point 182° to 187°C
Fieser 15,160; 16,170
CAS Max % 100.0
CAS Min % 94.0
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
Chemical Name or Material N-Fluoropyridinium triflate
Name Note 95%
Molecular Formula C6H5F4NO3S
CAS 107263-95-6
MDL Number MFCD00013458

C-X bond formation reagent

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.