Please login to your online account to display your discounted pricing

N-Fluoropyridinium triflate, 95%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  259925000

 View more versions of this product

Catalog No. AC259925000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystalline Powder or Needles
Formula Weight 247.16
Quantity 500mg
Color White to off-white
Melting Point 182° to 187°C
Fieser 15,160; 16,170
CAS Max % 100.0
CAS Min % 94.0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Packaging Glass bottle
Chemical Name or Material N-Fluoropyridinium triflate
Infrared Spectrum Authentic
Assay Percent Range 95%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 95%
Molecular Formula C6H5F4NO3S
MDL Number MFCD00013458
CAS 107263-95-6

Specifications

Chemical Name or Material N-Fluoropyridinium triflate
Name Note 95%
CAS 107263-95-6
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C6H5F4NO3S
Formula Weight 247.16
MDL Number MFCD00013458
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents