missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Fluorobenzenesulfonimide [Fluorinating Reagent], TCI America™

Manufacturer:  TCI America F033525G

 View more versions of this product

Catalog No. F033525G



Specifications

Specifications

N-Fluorobenzenesulfonimide [Fluorinating Reagent]
115°C
133745-75-2
MFCD00144885
RLKHFSNWQCZBDC-UHFFFAOYSA-N
N-(benzenesulfonyl)-N-fluorobenzenesulfonamide
588007
Crystal-Powder at 20°C
White-Yellow
25g
C12H10FNO4S2
n-fluorobenzenesulfonimide, nfsi, n-fluoro-n-phenylsulfonyl benzenesulfonamide, n-fluorodibenzenesulfonimide, n-fluorobenzensulfonimide, n-fluorobenzenesulphonimide, accufluor nfsi, n-fluorodi benzenesulfonyl amine, bis phenylsulfonyl fluoroamine, n-fluorobis phenylsulfonyl amine
C1=CC=C(C=C1)S(=O)(=O)N(F)S(=O)(=O)C2=CC=CC=C2
315.333
315.33
≥97.0% (T)

Chemical Identifiers

133745-75-2
315.333
RLKHFSNWQCZBDC-UHFFFAOYSA-N
588007
C1=CC=C(C=C1)S(=O)(=O)N(F)S(=O)(=O)C2=CC=CC=C2
C12H10FNO4S2
MFCD00144885
n-fluorobenzenesulfonimide, nfsi, n-fluoro-n-phenylsulfonyl benzenesulfonamide, n-fluorodibenzenesulfonimide, n-fluorobenzensulfonimide, n-fluorobenzenesulphonimide, accufluor nfsi, n-fluorodi benzenesulfonyl amine, bis phenylsulfonyl fluoroamine, n-fluorobis phenylsulfonyl amine
N-(benzenesulfonyl)-N-fluorobenzenesulfonamide
Safety and Handling

Safety and Handling

TSCA : No

Documents