missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Fluorobenzenesulfonimide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1395514

 View more versions of this product

Catalog No. AAL1395514



Description

Description

N-Fluorobenzenesulfonimide fluorinates aromatics, enolates, azaenolates and carbanions in high yield. It is used for diastereoselective fluorination of Li enolates of chiral carboximides, for directed fluorination of ortho-lithiated aromatics and for a review of electrophilic N-F fluorinating agents. Asymmteric fluorinations is effected in the presence of a Pd-BINAP catalyst system at room temperature in an ionic liquid. It is a reagent employed in a palladium-catalyzed enantioselective fluorination of t-butoxycarbonyl lactones and lactams. It is also used in the electrophilic difluorination of dihalopyridines with butyl lithium and in the direct conversion of alcohols to dibenzenesulfonamides with triphenylphosphine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fluorobenzenesulfonimide fluorinates aromatics, enolates, azaenolates and carbanions in high yield. It is used for diastereoselective fluorination of Li enolates of chiral carboximides, for directed fluorination of ortho-lithiated aromatics and for a review of electrophilic N-F fluorinating agents. Asymmteric fluorinations is effected in the presence of a Pd-BINAP catalyst system at room temperature in an ionic liquid. It is a reagent employed in a palladium-catalyzed enantioselective fluorination of t-butoxycarbonyl lactones and lactams. It is also used in the electrophilic difluorination of dihalopyridines with butyl lithium and in the direct conversion of alcohols to dibenzenesulfonamides with triphenylphosphine.

Solubility
Very soluble in acetonitrile, dichloromethane or THF and less soluble in toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Specifications

N-Fluorobenzenesulfonimide
114°C to 118°C
MFCD00144885
5348902
Very soluble in acetonitrile,dichloromethane or THF and less soluble in toluene.
C1=CC=C(C=C1)S(=O)(=O)N(F)S(=O)(=O)C2=CC=CC=C2
315.333
315.34
133745-75-2
C12H10FNO4S2
25 g
n-fluorobenzenesulfonimide, nfsi, n-fluoro-n-phenylsulfonyl benzenesulfonamide, n-fluorodibenzenesulfonimide, n-fluorobenzensulfonimide, n-fluorobenzenesulphonimide, accufluor nfsi, n-fluorodi benzenesulfonyl amine, bis phenylsulfonyl fluoroamine, n-fluorobis phenylsulfonyl amine
RLKHFSNWQCZBDC-UHFFFAOYSA-N
N-(benzenesulfonyl)-N-fluorobenzenesulfonamide
588007
97%

Chemical Identifiers

133745-75-2
315.333
RLKHFSNWQCZBDC-UHFFFAOYSA-N
588007
C1=CC=C(C=C1)S(=O)(=O)N(F)S(=O)(=O)C2=CC=CC=C2
C12H10FNO4S2
MFCD00144885
n-fluorobenzenesulfonimide, nfsi, n-fluoro-n-phenylsulfonyl benzenesulfonamide, n-fluorodibenzenesulfonimide, n-fluorobenzensulfonimide, n-fluorobenzenesulphonimide, accufluor nfsi, n-fluorodi benzenesulfonyl amine, bis phenylsulfonyl fluoroamine, n-fluorobis phenylsulfonyl amine
N-(benzenesulfonyl)-N-fluorobenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only