Please login to your online account to display your discounted pricing

N-Fluoro-2,6-dichloropyridinium triflate, 97%, Acros Organics


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC383270010*

 View more versions of this product

Catalog No. AC383270010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 316.06
Quantity 1g
Color Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
MDL Number MFCD00191438
CAS 130433-68-0
Health Hazard 1 Danger
Chemical Name or Material N-Fluoro-2, 6-dichloropyridinium triflate
Molecular Formula C6H3Cl2F4NO3S
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material N-Fluoro-2, 6-dichloropyridinium triflate
CAS 130433-68-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H3Cl2F4NO3S
Formula Weight 316.06
MDL Number MFCD00191438
Accessories

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes severe skin burns and eye damage.

SDS & Safety Documents