Please login to your online account to display your discounted pricing

N-Fluoro-2,6-dichloropyridinium triflate, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material N-Fluoro-2, 6-dichloropyridinium triflate
CAS 130433-68-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H3Cl2F4NO3S
Formula Weight 316.06
MDL Number MFCD00191438
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC383270010

  SDS
thermo fisher scientific
AC383270010*
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder
Formula Weight 316.06
Quantity 1g
Color Yellow
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 1 Danger
Assay Percent Range 97%
MDL Number MFCD00191438
CAS 130433-68-0
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C6H3Cl2F4NO3S
Chemical Name or Material N-Fluoro-2, 6-dichloropyridinium triflate
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.

C-X bond formation reagent

Safety and Handling

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes severe skin burns and eye damage.