missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Ethyltoluenesulfonamide (o- and p- mixture), TCI America™

Manufacturer:  TCI America E0187500G

 View more versions of this product

Catalog No. E0187500G



Specifications

Specifications

N-Ethyltoluenesulfonamide (o- and p- mixture)
500g
C18H26N2O4S2
IDLSDSKPHLCUTN-UHFFFAOYSA-N
199.27
≥98.0% (N)
Yellow
76902-32-4
MFCD00048511
398.54
Clear Liquid at 20°C

Chemical Identifiers

76902-32-4
398.54
IDLSDSKPHLCUTN-UHFFFAOYSA-N
C18H26N2O4S2
MFCD00048511
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1929

TSCA : No

Documents