missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethylpropylamine, 97+%

Manufacturer:  Alfa Aesar™ 1484606

 View more versions of this product

Catalog No. AA1484606



Specifications

Specifications

N-Ethylpropylamine
4.4° (40°F)
5g
C5H13N
UN2733
XCVNDBIXFPGMIW-UHFFFAOYSA-N
N-ethylpropan-1-amine
88398
Liquid
0.748g/mL
80°C to 85°C
20193-20-8
MFCD00015198
n-ethylpropylamine, 1-propanamine, n-ethyl, ethyl propyl amine, propylamine, n-ethyl, ethylpropylamine, n-ethyl-n-propylamine, ethyl-propyl-amine, n-propylethylamine, n-ethylpropanamine, ethyl-n-propylamine
CCCNCC
87.166
87.16
≥97%

Chemical Identifiers

20193-20-8
87.166
XCVNDBIXFPGMIW-UHFFFAOYSA-N
88398
CCCNCC
C5H13N
MFCD00015198
n-ethylpropylamine, 1-propanamine, n-ethyl, ethyl propyl amine, propylamine, n-ethyl, ethylpropylamine, n-ethyl-n-propylamine, ethyl-propyl-amine, n-propylethylamine, n-ethylpropanamine, ethyl-n-propylamine
N-ethylpropan-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

TSCA : TSCA

SDS Documents