Learn More
N-Ethylmorpholine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 149405000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Ethylmorpholine | |
100-74-3 | |
100.0 | |
Brown-Gray to Cream | |
139.0°C | |
Authentic | |
Glass bottle | |
1.4400 to 1.4420 | |
500 mL | |
01,383 | |
n-ethylmorpholine, morpholine, 4-ethyl, ethylmorpholine, n-ethylmorfolin, n-ethyl morpholine, n-ethylmorfolin czech, unii-ecm0g991fq, n-ethyl-morpholine, ccris 4818, ecm0g991fq | |
HVCNXQOWACZAFN-UHFFFAOYSA-N | |
4-ethylmorpholine | |
7525 | |
99% |
99% | |
98.5 | |
-63.0°C | |
0.9000g/mL | |
27°C | |
98.5% min. (GC) | |
C6H13NO | |
MFCD00006177 | |
27, 6 | |
0.9 | |
Solubility in water: miscible with water. Other solubilities: soluble in alcohol,ether,acetone and benzene | |
CCN1CCOCC1 | |
115.17 | |
115.17 | |
Powder |
Chemical Identifiers
100-74-3 | |
115.17 | |
HVCNXQOWACZAFN-UHFFFAOYSA-N | |
7525 | |
CCN1CCOCC1 |
C6H13NO | |
MFCD00006177 | |
n-ethylmorpholine, morpholine, 4-ethyl, ethylmorpholine, n-ethylmorfolin, n-ethyl morpholine, n-ethylmorfolin czech, unii-ecm0g991fq, n-ethyl-morpholine, ccris 4818, ecm0g991fq | |
4-ethylmorpholine |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 202-885-0
RTECSNumber : QE4025000
TSCA : TSCA
RUO – Research Use Only