missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethylmethylamine, 98+%

Manufacturer:  Alfa Aesar™ L0437214

 View more versions of this product

Catalog No. AAL0437214



Specifications

Specifications

N-Ethylmethylamine
Amine-like
25g
C3H9N
UN2733
ethanamine, n-methyl
CCNC
59.112
59.11
<−34°C (−29°F)
33°C to 34°C
624-78-2
MFCD00009030
1730786
LIWAQLJGPBVORC-UHFFFAOYSA-N
N-methylethanamine
12219
≥98%

Chemical Identifiers

624-78-2
59.112
LIWAQLJGPBVORC-UHFFFAOYSA-N
12219
CCNC
C3H9N
MFCD00009030
ethanamine, n-methyl
N-methylethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H224-H314-H318
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents