missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Ethylmaleimide, TCI America™

Manufacturer:  TCI America E01365G

 View more versions of this product

Catalog No. E01365G



Specifications

Specifications

N-Ethylmaleimide
White
5g
C6H7NO2
2928
HDFGOPSGAURCEO-UHFFFAOYSA-N
1-ethylpyrrole-2,5-dione
4362
125.13
≥98.0% (HPLC,N)
210°C
46°C
128-53-0
MFCD00005509
n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide
CCN1C(=O)C=CC1=O
125.127
CHEBI:44485
Crystal-Powder at 20°C

Chemical Identifiers

128-53-0
125.127
HDFGOPSGAURCEO-UHFFFAOYSA-N
4362
1-ethylpyrrole-2,5-dione
C6H7NO2
MFCD00005509
n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide
CHEBI:44485
CCN1C(=O)C=CC1=O
Safety and Handling

Safety and Handling

RTECSNumber : UX9625000

TSCA : Yes

Recommended Storage : Refrigerator

Documents