missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Ethylmaleimide, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC156102500

 View more versions of this product

Catalog No. AC156102500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-Ethylmaleimide
Authentic
210.0°C
41.0°C to 45.0°C
128-53-0
C6H7NO2
15,3877
Solubility in water: soluble. Other solubilities: 1115g/L toluene (20°C)
CCN1C(=O)C=CC1=O
125.13
CHEBI:44485
Crystalline Powder
73°C
Glass Bottle
White
250g
99% min. (GC)
21,IV,4629
1-ethyl-1h-pyrrole-2,5-dione
HDFGOPSGAURCEO-UHFFFAOYSA-N
1-ethylpyrrole-2,5-dione
4362
125.13
99+%

Chemical Identifiers

128-53-0
125.13
1-ethyl-1h-pyrrole-2,5-dione
CHEBI:44485
CCN1C(=O)C=CC1=O
C6H7NO2
HDFGOPSGAURCEO-UHFFFAOYSA-N
4362
1-ethylpyrrole-2,5-dione
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 204-892-4

SDS Documents