missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethylmaleamic acid, tech. 85%

$91.14 - $300.72

Chemical Identifiers

CAS 4166-67-0
Molecular Formula C6H9NO3
Molecular Weight (g/mol) 143.142
MDL Number MFCD00014013
InChI Key HBQGCOWNLUOCBU-ARJAWSKDSA-N
Synonym n-ethylmaleamic acid, 2z-3-ethylcarbamoyl prop-2-enoic acid, 2z-3-n-ethylcarbamoyl prop-2-enoic acid, n-ethylmaleamidic acid, n-ethyl-maleamic acid, z-4-ethylamino-4-oxobut-2-enoic acid, 2z-4-ethylamino-4-oxo-2-butenoic acid, 2z-4-ethylamino-4-oxobut-2-enoic acid
PubChem CID 5380356
IUPAC Name (Z)-4-(ethylamino)-4-oxobut-2-enoic acid
SMILES CCNC(=O)C=CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0064509 View Documents Alfa Aesar™
L0064509
10g Each for $91.14
 
AAL0064518 View Documents Alfa Aesar™
L0064518
50g Each for $300.72
 
Specifications

Chemical Identifiers

4166-67-0
143.142
HBQGCOWNLUOCBU-ARJAWSKDSA-N
5380356
CCNC(=O)C=CC(=O)O
C6H9NO3
MFCD00014013
n-ethylmaleamic acid, 2z-3-ethylcarbamoyl prop-2-enoic acid, 2z-3-n-ethylcarbamoyl prop-2-enoic acid, n-ethylmaleamidic acid, n-ethyl-maleamic acid, z-4-ethylamino-4-oxobut-2-enoic acid, 2z-4-ethylamino-4-oxo-2-butenoic acid, 2z-4-ethylamino-4-oxobut-2-enoic acid
(Z)-4-(ethylamino)-4-oxobut-2-enoic acid

Specifications

124°C to 126°C
4166-67-0
MFCD00014013
n-ethylmaleamic acid, 2z-3-ethylcarbamoyl prop-2-enoic acid, 2z-3-n-ethylcarbamoyl prop-2-enoic acid, n-ethylmaleamidic acid, n-ethyl-maleamic acid, z-4-ethylamino-4-oxobut-2-enoic acid, 2z-4-ethylamino-4-oxo-2-butenoic acid, 2z-4-ethylamino-4-oxobut-2-enoic acid
CCNC(=O)C=CC(=O)O
143.142
143.14
Technical
10g
C6H9NO3
1210030
HBQGCOWNLUOCBU-ARJAWSKDSA-N
(Z)-4-(ethylamino)-4-oxobut-2-enoic acid
5380356
85%
N-Ethylmaleamic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA