Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Ethylheptylamine, 97%

Chemical Identifiers

Chemical Name or Material N-Ethylheptylamine
CAS 66793-76-8
Linear Formula CH3CH2NH(CH2)6CH3
Formula Weight 143.27
MDL Number MFCD00049012
UN Number UN2733
Beilstein 1733571
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6510703

  SDS
alfa aesar™
H6510703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6510706

  SDS
alfa aesar™
H6510706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6510714

  SDS
alfa aesar™
H6510714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 143.27
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
CAS 66793-76-8
MDL Number MFCD00049012
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material N-Ethylheptylamine
Beilstein 1733571
UN Number UN2733
Color Pale yellow
Physical Form Liquid
Assay 97%
Refractive Index 1.4240
Boiling Point 181°C
Quantity 1g
Linear Formula CH3CH2NH(CH2)6CH3
Density 0.772g/mL
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.