missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Ethylglycine 98.0+%, TCI America™
Supplier: TCI America E051725G
Specifications
N-Ethylglycine | |
190°C | |
C4H9NO2 | |
25 g | |
YPIGGYHFMKJNKV-UHFFFAOYSA-N | |
2-(ethylamino)acetic acid | |
316542 | |
103.12 | |
Crystalline Powder |
627-01-0 | |
White-Yellow | |
MFCD00037794 | |
n-ethylglycine, 2-ethylamino acetic acid, n-ethyl glycine, etgly, glycine, n-ethyl, monoethylglycine, pubchem23122, acmc-1au6b, ypiggyhfmkjnkv-uhfffaoysa | |
CCNCC(=O)O | |
103.121 | |
CHEBI:15620 | |
≥98.0% (T) |
Chemical Identifiers
627-01-0 | |
103.121 | |
YPIGGYHFMKJNKV-UHFFFAOYSA-N | |
316542 | |
2-(ethylamino)acetic acid |
C4H9NO2 | |
MFCD00037794 | |
n-ethylglycine, 2-ethylamino acetic acid, n-ethyl glycine, etgly, glycine, n-ethyl, monoethylglycine, pubchem23122, acmc-1au6b, ypiggyhfmkjnkv-uhfffaoysa | |
CHEBI:15620 | |
CCNCC(=O)O |
Safety and Handling
TSCA : No