Please login to your online account to display your discounted pricing

N-ethylglycine 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Ethylglycine
CAS 627-01-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H9NO2
Linear Formula CH3CH2NHCH2COOH
Formula Weight 103.12
MDL Number MFCD00037794
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC381420010

  SDS
thermo fisher scientific
AC381420010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 103.12
Physical Form Powder
Quantity 1g
Color White to almost white
Melting Point 182° to 186°C
CAS Max % 100.0
CAS 627-01-0
Assay Percent Range 98%
Molecular Formula C4H9NO2
Chemical Name or Material N-Ethylglycine
MDL Number MFCD00037794
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula CH3CH2NHCH2COOH