missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Ethylethylenediamine, TCI America™

Manufacturer:  TCI America E029425ML

 View more versions of this product

Catalog No. E029425ML



Specifications

Specifications

N-Ethylethylenediamine
Yellow
110-72-5
MFCD00008166
n-ethylethylenediamine, 1,2-ethanediamine, n-ethyl, 2-aminoethyl ethyl amine, 2-ethylaminoethylamine, ethylenediamine, n-ethyl, ethylaminoethylamine, unii-941mvd708n, 1,2-ethanediamine, n1-ethyl, n-ethyl ethylenediamine, 2-aminoethyl ethyl amine
CCNCCN
88.154
88.15
≥99.0% (GC)
130°C
25mL
C4H12N2
2734
SCZVXVGZMZRGRU-UHFFFAOYSA-N
N'-ethylethane-1,2-diamine
66071
Clear Liquid at 20°C

Chemical Identifiers

110-72-5
88.154
SCZVXVGZMZRGRU-UHFFFAOYSA-N
66071
CCNCCN
C4H12N2
MFCD00008166
n-ethylethylenediamine, 1,2-ethanediamine, n-ethyl, 2-aminoethyl ethyl amine, 2-ethylaminoethylamine, ethylenediamine, n-ethyl, ethylaminoethylamine, unii-941mvd708n, 1,2-ethanediamine, n1-ethyl, n-ethyl ethylenediamine, 2-aminoethyl ethyl amine
N'-ethylethane-1,2-diamine
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3275

TSCA : Yes

Documents