Please login to your online account to display your discounted pricing

N-Ethylethylenediamine, 98+%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  155552500

 View more versions of this product

Catalog No. AC155552500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Density 0.8300g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4380 to 1.4400 (20°C, 589nm)
Color Clear colorless to slightly brown
Quantity 250g
Chemical Name or Material N-Ethylethylenediamine
CAS 110-72-5
CAS Min % 98.0
CAS Max % 100.0
Physical Form Clear colorless to slightly brown liquid
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C4H12N2
Linear Formula C2H5NHCH2CH2NH2
Formula Weight 88.15g/mol
MDL Number MFCD00008166
Beilstein 04, II, 690
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material N-Ethylethylenediamine
CAS 110-72-5
CAS Min % 98.0
CAS Max % 100.0
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C4H12N2
Linear Formula C2H5NHCH2CH2NH2
Formula Weight 88.15g/mol
MDL Number MFCD00008166
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

SDS & Safety Documents