missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Ethyldiisopropylamine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A11801AE

 View more versions of this product

Catalog No. AAA11801AE


Description

Description

N-Ethyldiisopropylamine is used to make or formulate industrial products. It is also used in organic chemistry as a base. It acts as a selective reagent in the alkylation of secondary amines to tertiary amines by alkyl halides. It reacts with monoactivated Michael acceptors to afford symmetrical sulfones. Further, it is used to prepare heterocyclic compound, sscorpionine by a reaction with disulfur dichloride using 1,4-diazabicyclo[2.2.2]octane (DABCO) as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Ethyldiisopropylamine is used to make or formulate industrial products. It is also used in organic chemistry as a base. It acts as a selective reagent in the alkylation of secondary amines to tertiary amines by alkyl halides. It reacts with monoactivated Michael acceptors to afford symmetrical sulfones. Further, it is used to prepare heterocyclic compound, sscorpionine by a reaction with disulfur dichloride using 1,4-diazabicyclo[2.2.2]octane (DABCO) as a catalyst.

Solubility
Miscible with water and ethanol.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, carbon dioxide, copper, brass, rubber, oxidizing agents, nitrates, peroxides, aldehydes and metals.
Specifications

Specifications

N-Ethyldiisopropylamine
−46°C
126°C to 127°C
99%
1.4135
100 mL
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
JGFZNNIVVJXRND-UHFFFAOYSA-N
N-ethyl-N-propan-2-ylpropan-2-amine
81531
Liquid
7087-68-5
0.747
8°C (46°F)
C8H19N
MFCD00008868
605301
Miscible with water and ethanol.
CCN(C(C)C)C(C)C
129.247
129.25

Chemical Identifiers

7087-68-5
129.247
JGFZNNIVVJXRND-UHFFFAOYSA-N
81531
CCN(C(C)C)C(C)C
C8H19N
MFCD00008868
n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
N-ethyl-N-propan-2-ylpropan-2-amine
Safety and Handling

Safety and Handling

P210-P233-P234-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P330-P370+P378q-P390-P501c

H225-H290-H302-H318-H331-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: I

EINECSNumber : 230-392-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only