Learn More
N-Ethylcyclohexylamine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 437740250
Specifications
N-Ethylcyclohexylamine | |
97.5 | |
0.8500g/mL | |
30°C | |
97.5% min. (GC) | |
C8H17N | |
25mL | |
n-ethylcyclohexylamine, cyclohexanamine, n-ethyl, n-cyclohexylethylamine, vulkacit hx, accelerator hx, cyclohexylamine, n-ethyl, ethylamino cyclohexane, cyclohexyl ethyl amine, n-ethyl cyclohexyl amine, unii-yjk13p0h3e | |
CCNC1CCCCC1 | |
127.23 | |
127.23 |
5459-93-8 | |
100.0 | |
164°C | |
Authentic | |
Glass bottle | |
1.4500 to 1.4530 (20°C, 589nm) | |
0.85 | |
AGVKXDPPPSLISR-UHFFFAOYSA-N | |
N-ethylcyclohexanamine | |
21609 | |
98% |
Chemical Identifiers
5459-93-8 | |
127.23 | |
n-ethylcyclohexylamine, cyclohexanamine, n-ethyl, n-cyclohexylethylamine, vulkacit hx, accelerator hx, cyclohexylamine, n-ethyl, ethylamino cyclohexane, cyclohexyl ethyl amine, n-ethyl cyclohexyl amine, unii-yjk13p0h3e | |
N-ethylcyclohexanamine |
C8H17N | |
AGVKXDPPPSLISR-UHFFFAOYSA-N | |
21609 | |
CCNC1CCCCC1 |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 226-733-8
RUO â Research Use Only