Please login to your online account to display your discounted pricing

N-Ethylcarbazole 98.5%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-Ethylcarbazole
Name Note 98.5%
CAS 86-28-2
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98.5%
Molecular Formula C14 H13 N
Formula Weight 195.26
MDL Number MFCD00004967
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC118215000

  SDS
thermo fisher scientific
118215000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Flakes
Assay ≥98.0%
Formula Weight 195.26
Quantity 500g
Color Brown
EINECS Number 201-660-4
TSCA TSCA
Melting Point 66° to 70°C
RTECS Number FE6225700
Beilstein 20, 436
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material N-Ethylcarbazole
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98.5%
Assay Percent Range 98.5%
Molecular Formula C14 H13 N
MDL Number MFCD00004967
CAS 86-28-2